Assay Details
Gene Symbol: Amdhd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024990.2 | NP_001020161.2 | 1 - 2 | 150 | 76 | |
XM_008767565.1 | XP_008765787.1 | 1 - 2 | 241 | 76 | ||
GenBank mRNA | BC091278.1 | - | 1 - 2 | 140 | 76 | |
CO388545.1 | - | 1 - 2 | 150 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
