Assay Details
Gene Symbol: Tlk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191652.1 | NP_001178581.1 | 13 - 14 | 1716 | 146 | |
XM_006247574.2 | XP_006247636.1 | 13 - 14 | 3059 | 146 | ||
XM_006247576.2 | XP_006247638.1 | 13 - 14 | 1612 | 146 | ||
XM_008768348.1 | XP_008766570.1 | 12 - 13 | 1577 | 146 | ||
XM_008768349.1 | XP_008766571.1 | 11 - 12 | 1485 | 146 | ||
XM_008768350.1 | XP_008766572.1 | 8 - 9 | 909 | 146 | ||
GenBank mRNA | BC086544.1 | - | 3 - 4 | 321 | 146 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
