Assay Details
Gene Symbol: Prpsap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057131.1 | NP_476472.1 | 1 - 2 | 87 | 132 | |
XM_006246439.2 | XP_006246501.1 | 1 - 2 | 169 | 131 | ||
XM_006246440.2 | XP_006246502.1 | 1 - 2 | 162 | 131 | ||
XM_008767764.1 | XP_008765986.1 | 1 - 2 | 160 | 131 | ||
GenBank mRNA | D84434.1 | - | 1 - 2 | 87 | 132 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the D84434.1, NM_057131.1, XM_006246439.2, XM_006246440.2, XM_008767764.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
