Assay Details
Gene Symbol: Ublcp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014117.1 | NP_001014139.1 | 1 - 2 | 33 | 110 | |
XM_006246140.1 | XP_006246202.1 | 1 - 2 | 93 | 110 | ||
GenBank mRNA | BC089210.1 | - | 1 - 2 | 33 | 7302690 | 110 |
FQ233702.1 | - | 1 - 2 | 66 | 110 | ||
FQ233841.1 | - | 1 - 2 | 66 | 110 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC089210.1,NM_001014117.1,XM_006246140.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
