Assay Details
Gene Symbol: Ublcp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014117.1 | NP_001014139.1 | 2 - 3 | 237 | 116 | |
XM_006246140.1 | XP_006246202.1 | 2 - 3 | 297 | 116 | ||
GenBank mRNA | BC089210.1 | - | 2 - 3 | 237 | 116 | |
FQ233702.1 | - | 2 - 3 | 270 | 116 | ||
FQ233841.1 | - | 2 - 3 | 270 | 116 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
