Assay Details
Gene Symbol: Ublcp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014117.1 | NP_001014139.1 | 5 - 6 | 529 | 76 | |
XM_006246140.1 | XP_006246202.1 | 5 - 6 | 559 | 76 | ||
GenBank mRNA | BC089210.1 | - | 5 - 6 | 529 | 7302690 | 76 |
FQ233702.1 | - | 5 - 6 | 532 | 76 | ||
FQ233841.1 | - | 5 - 6 | 532 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
