Assay Details
Gene Symbol: Smg6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105808.1 | NP_001099278.1 | 1 - 2 | 668 | 74 | |
XM_006246866.2 | XP_006246928.1 | 1510 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
