Assay Details
Gene Symbol: Rad51c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129777.1 | NP_001123249.1 | 2 - 3 | 409 | 84 | |
XM_006247106.1 | XP_006247168.1 | 2 - 3 | 487 | 84 | ||
GenBank mRNA | BC166862.1 | - | 2 - 3 | 316 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
