Assay Details
Gene Symbol: Rad51c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129777.1 | NP_001123249.1 | 4 - 5 | 703 | 96 | |
XM_006247106.1 | XP_006247168.1 | 4 - 5 | 781 | 96 | ||
GenBank mRNA | BC166862.1 | - | 4 - 5 | 610 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
