Assay Details
Gene Symbol: Alox15b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153301.2 | NP_695213.1 | 10 - 11 | 1494 | 91 | |
XM_008767779.1 | XP_008766001.1 | 9 - 10 | 1355 | 91 | ||
GenBank mRNA | AF415240.1 | - | 10 - 11 | 1502 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
