Assay Details
Gene Symbol: Alox15b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153301.2 | NP_695213.1 | 12 - 13 | 1708 | 107 | |
XM_008767779.1 | XP_008766001.1 | 11 - 12 | 1569 | 107 | ||
GenBank mRNA | AF415240.1 | - | 12 - 13 | 1716 | 107 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
