Assay Details
Gene Symbol: Alox15b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153301.2 | NP_695213.1 | 7 - 8 | 1024 | 54 | |
XM_008767779.1 | XP_008766001.1 | 6 - 7 | 885 | 54 | ||
GenBank mRNA | AF415240.1 | - | 7 - 8 | 1032 | 54 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
