Assay Details
Gene Symbol: Sap30bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108305.1 | NP_001101775.1 | 2 - 3 | 250 | 78 | |
GenBank mRNA | BC169106.1 | - | 219 | 78 | ||
FQ220147.1 | - | 2 - 3 | 247 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
