Assay Details
Gene Symbol: Sap30bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108305.1 | NP_001101775.1 | 5 - 6 | 439 | 94 | |
GenBank mRNA | BC169106.1 | - | 408 | 94 | ||
FQ220147.1 | - | 5 - 6 | 436 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
