Assay Details
Gene Symbol: Sap30bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108305.1 | NP_001101775.1 | 7 - 8 | 579 | 122 | |
GenBank mRNA | BC169106.1 | - | 548 | 122 | ||
FQ220147.1 | - | 7 - 8 | 576 | 122 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
