Assay Details
Gene Symbol: C1qtnf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191918.1 | NP_001178847.1 | 2 - 3 | 142 | 84 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001191918.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
