Assay Details
Gene Symbol: Dhrs7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008507.1 | NP_001008507.1 | 4 - 5 | 536 | 67 | |
XM_006246472.2 | XP_006246534.1 | 5 - 6 | 744 | 67 | ||
XM_006246473.2 | XP_006246535.1 | 4 - 5 | 1405 | 67 | ||
XM_006246474.2 | XP_006246536.1 | 4 - 5 | 873 | 67 | ||
XM_008767786.1 | XP_008766008.1 | 4 - 5 | 732 | 67 | ||
GenBank mRNA | BC086453.1 | - | 4 - 5 | 536 | 67 | |
EF445633.1 | - | 5 - 6 | 648 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
