Assay Details
Gene Symbol: Alox15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031010.2 | NP_112272.2 | 8 - 9 | 1171 | 140 | |
XM_006246823.2 | XP_006246885.1 | 10 - 11 | 1452 | 140 | ||
XM_006246824.2 | XP_006246886.1 | 9 - 10 | 1347 | 140 | ||
GenBank mRNA | FQ233819.1 | - | 2 - 3 | 299 | 140 | |
L06040.1 | - | 8 - 9 | 1184 | 140 | ||
S69383.1 | - | 8 - 9 | 1171 | 140 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
