Assay Details
Gene Symbol: Arhgef15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105789.1 | NP_001099259.1 | 1 - 2 | 70 | 89 | |
XM_008767788.1 | XP_008766010.1 | 1 - 2 | 1316 | 89 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001105789.1, XM_008767788.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
