Assay Details
Gene Symbol: Map2k6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053703.3 | NP_446155.1 | 1 - 2 | 129 | 139 | |
GenBank mRNA | AF369384.1 | - | 1 - 2 | 129 | 139 | |
BC087004.1 | - | 1 - 2 | 229 | 139 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AF369384.1, BC087004.1, NM_053703.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
