Assay Details
Gene Symbol: Nat9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134835.1 | NP_001128307.1 | 3 - 4 | 753 | 129 | |
XM_006247714.2 | XP_006247776.1 | 4 - 5 | 612 | 129 | ||
GenBank mRNA | BC158710.1 | - | 3 - 4 | 753 | 129 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
