Assay Details
Gene Symbol: Ttc25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107049.1 | NP_001100519.1 | 3 - 4 | 524 | 66 | |
XM_006247310.2 | XP_006247372.1 | 3 - 4 | 1378 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
