Assay Details
Gene Symbol: Cdc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108298.1 | NP_001101768.1 | 6 - 7 | 1127 | 66 | |
XM_006247354.2 | XP_006247416.1 | 6 - 7 | 1129 | 66 | ||
XM_006247355.2 | XP_006247417.1 | 6 - 7 | 1127 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
