Assay Details
Gene Symbol: Kcnh6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053937.1 | NP_446389.1 | 10 - 11 | 2266 | 66 | |
XM_006247554.2 | XP_006247616.1 | 11 - 12 | 2357 | 66 | ||
XM_006247555.2 | XP_006247617.1 | 11 - 12 | 2315 | 66 | ||
XM_006247556.2 | XP_006247618.1 | 10 - 11 | 2294 | 66 | ||
XM_008768308.1 | XP_008766530.1 | 11 - 12 | 2361 | 66 | ||
XM_008768309.1 | XP_008766531.1 | 10 - 11 | 2290 | 66 | ||
GenBank mRNA | AF016192.1 | - | 10 - 11 | 2266 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
