Assay Details
Gene Symbol: Tp53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030989.3 | NP_112251.2 | 2 - 3 | 207 | 134 | |
XM_008767773.1 | XP_008765995.1 | 2 - 3 | 214 | 134 | ||
GenBank mRNA | BC081788.1 | - | 2 - 3 | 136 | 134 | |
BC098663.1 | - | 2 - 3 | 165 | 134 | ||
CV126174.1 | - | 2 - 3 | 207 | 134 | ||
FQ232659.1 | - | 2 - 3 | 191 | 134 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC081788.1, BC098663.1, NM_030989.3, XM_008767773.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
