Assay Details
Gene Symbol: Tp53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030989.3 | NP_112251.2 | 3 - 4 | 232 | 112 | |
XM_006246594.2 | XP_006246656.1 | 4 - 5 | 661 | 112 | ||
XM_006246595.2 | XP_006246657.1 | 3 - 4 | 1018 | 112 | ||
XM_008767773.1 | XP_008765995.1 | 3 - 4 | 239 | 112 | ||
GenBank mRNA | AF209191.1 | - | 2 - 3 | 124 | 112 | |
AY009504.1 | - | 2 - 3 | 101 | 112 | ||
BC081788.1 | - | 3 - 4 | 161 | 7193583 | 112 | |
BC098663.1 | - | 3 - 4 | 190 | 7384467 | 112 | |
CV126174.1 | - | 3 - 4 | 232 | 112 | ||
U90328.1 | - | 2 - 3 | 101 | 112 | ||
X13058.1 | - | 2 - 3 | 124 | 112 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
