Assay Details
Gene Symbol: Tp53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030989.3 | NP_112251.2 | 4 - 5 | 501 | 90 | |
XM_006246594.2 | XP_006246656.1 | 5 - 6 | 930 | 90 | ||
XM_006246595.2 | XP_006246657.1 | 4 - 5 | 1287 | 90 | ||
XM_008767773.1 | XP_008765995.1 | 4 - 5 | 508 | 90 | ||
GenBank mRNA | AF209191.1 | - | 3 - 4 | 393 | 90 | |
BC081788.1 | - | 4 - 5 | 430 | 90 | ||
BC098663.1 | - | 4 - 5 | 459 | 90 | ||
CV126174.1 | - | 4 - 5 | 501 | 90 | ||
FQ232659.1 | - | 4 - 5 | 485 | 90 | ||
U90328.1 | - | 3 - 4 | 370 | 90 | ||
X13058.1 | - | 3 - 4 | 393 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
