Assay Details
Gene Symbol: B4galnt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008768250.1 | XP_008766472.1 | 5 - 6 | 875 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
