Assay Details
Gene Symbol: Apoh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009626.1 | NP_001009626.1 | 6 - 7 | 788 | 77 | |
GenBank mRNA | AM071386.1 | - | 6 - 7 | 747 | 77 | |
BC088180.1 | - | 6 - 7 | 788 | 7364928 | 77 | |
FQ209412.1 | - | 6 - 7 | 827 | 77 | ||
FQ209491.1 | - | 6 - 7 | 814 | 77 | ||
FQ209740.1 | - | 6 - 7 | 824 | 77 | ||
FQ210311.1 | - | 6 - 7 | 814 | 77 | ||
FQ210813.1 | - | 6 - 7 | 814 | 77 | ||
FQ211328.1 | - | 6 - 7 | 787 | 77 | ||
FQ218068.1 | - | 1 - 2 | 101 | 77 | ||
FQ218335.1 | - | 2 - 3 | 321 | 77 | ||
FQ218445.1 | - | 6 - 7 | 828 | 77 | ||
FQ218454.1 | - | 6 - 7 | 815 | 77 | ||
FQ218597.1 | - | 6 - 7 | 810 | 77 | ||
FQ218599.1 | - | 6 - 7 | 814 | 77 | ||
FQ218637.1 | - | 6 - 7 | 814 | 77 | ||
FQ218859.1 | - | 6 - 7 | 787 | 77 | ||
FQ218865.1 | - | 6 - 7 | 814 | 77 | ||
FQ218961.1 | - | 6 - 7 | 814 | 77 | ||
FQ219138.1 | - | 6 - 7 | 879 | 77 | ||
FQ219328.1 | - | 6 - 7 | 814 | 77 | ||
FQ219438.1 | - | 6 - 7 | 813 | 77 | ||
FQ219481.1 | - | 6 - 7 | 826 | 77 | ||
FQ219537.1 | - | 6 - 7 | 811 | 77 | ||
FQ219601.1 | - | 6 - 7 | 827 | 77 | ||
FQ219674.1 | - | 6 - 7 | 814 | 77 | ||
X15551.1 | - | 6 - 7 | 643 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
