Assay Details
Gene Symbol: Pdk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030872.1 | NP_110499.1 | 3 - 4 | 436 | 89 | |
GenBank mRNA | AF237719.1 | - | 3 - 4 | 436 | 89 | |
BC061823.1 | - | 3 - 4 | 400 | 89 | ||
U10357.1 | - | 3 - 4 | 436 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
