Assay Details
Gene Symbol: Gfra2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012750.1 | NP_036882.1 | 2 - 3 | 475 | 131 | |
XM_006252238.2 | XP_006252300.1 | 1 - 2 | 1244 | 131 | ||
GenBank mRNA | AF003825.1 | - | 1025 | 131 | ||
AF005226.1 | - | 2 - 3 | 355 | 131 | ||
U97143.1 | - | 2 - 3 | 475 | 131 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
