Assay Details
Gene Symbol: Gfra2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012750.1 | NP_036882.1 | 5 - 6 | 1021 | 63 | |
XM_006252238.2 | XP_006252300.1 | 4 - 5 | 1790 | 63 | ||
GenBank mRNA | AF003825.1 | - | 1571 | 63 | ||
AF005226.1 | - | 5 - 6 | 901 | 63 | ||
U97143.1 | - | 5 - 6 | 1021 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
