Assay Details
Gene Symbol: Gfra2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012750.1 | NP_036882.1 | 8 - 9 | 1399 | 134 | |
XM_006252238.2 | XP_006252300.1 | 7 - 8 | 2168 | 134 | ||
GenBank mRNA | AF003825.1 | - | 1949 | 134 | ||
AF005226.1 | - | 8 - 9 | 1279 | 134 | ||
U97143.1 | - | 8 - 9 | 1399 | 134 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P791436 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P146388 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P007628 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 972928 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 638448 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 325886 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 495432 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 478255 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 52299 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 516492 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 117781 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 792100 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 401927 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 318864 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 204395 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 235435 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 179185 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 891647 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 129420 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 611708 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 416875 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 167264 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 372793 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 43162 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 809746 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 737985 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 744855 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 220933 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 648401 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 968171 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 481885 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 723128 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 368783 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 479640 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 733328 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 820186 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 59609 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 537078 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 154391 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 633246 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 712956 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 479214 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 627138 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 217044 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 301201 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 398640 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 673407 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 522242 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 484702 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 524116 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 58265 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 721539 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 2704 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 520655 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 716870 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 123273 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 47856 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 915150 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 842502 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 392455 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 182572 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 802453 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 809786 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 110039 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 495475 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 916265 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 804797 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 67415 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 752410 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 883742 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 117000 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 234128 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 113402 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 18522 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 120860 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 83483 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 107159 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 71965 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 94925 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 41244 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 44374 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 164661 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 32023 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 84302 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 22414 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 102010 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 43429 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 46549 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P007628 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P791436 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P146388 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P146388 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P007628 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P791436 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P007628 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P791436 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P146388 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P791436 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 701912125 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 92724 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 104106 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 63664 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 42410 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 3609 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 75048 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 701912125 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10781413 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10781416 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10781418 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10781420 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10781423 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10781422 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10781417 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10781415 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10781412 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10781419 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10781421 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10781411 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10781414 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10781410 | - | - |
Rat Genome 230 2.0 Array | 1369167_PM_at | - | - |
Rat Genome 230 2.0 Array | 1375359_at | - | - |
Rat Genome 230 2.0 Array | 1375359_PM_at | - | - |
Rat Genome 230 2.0 Array | 1369167_at | - | - |
Affymetrix HT Rat Focus Array | 1369167_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1375359_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1375359_at | - | - |
Affymetrix HT Rat Focus Array | 1369167_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1500005514.rn.1 | TC1500000790.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1500005515.rn.1 | TC1500000790.rn.1 | RTA TC |
Rat Expression Set 230 | 1369167_PM_at | - | - |
Rat Expression Set 230 | 1369167_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10781410 | - | - |
Rat Genome U34 Set | AF003825_s_at | - | - |
Rat Genome U34 Set | U97143_at | - | - |
Rat Genome U34 Set | U97143_at | - | - |
Rat Genome U34 Set | AF003825_s_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21398174 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_2039843 | - | - |
RatRef-12 Expression BeadChip | ILMN_52069 | - | - |
