Assay Details
Gene Symbol: Polr3d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031653.1 | NP_001026823.1 | 2 - 3 | 210 | 99 | |
XM_006252272.2 | XP_006252334.1 | 2 - 3 | 210 | 99 | ||
GenBank mRNA | BC099167.1 | - | 2 - 3 | 210 | 99 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
