Assay Details
Gene Symbol: Stk24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127494.1 | NP_001120966.1 | 1055 | 88 | ||
GenBank mRNA | EU371958.1 | - | 1055 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
