Assay Details
Gene Symbol: Gpr180 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006252452.2 | XP_006252514.1 | 1 - 2 | 251 | 74 | |
XM_006252455.2 | XP_006252517.1 | 1 - 2 | 252 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
