Assay Details
Gene Symbol: Gpr180 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006994.1 | NP_001006995.1 | 2 - 3 | 315 | 64 | |
XM_006252452.2 | XP_006252514.1 | 2 - 3 | 411 | 64 | ||
XM_006252455.2 | XP_006252517.1 | 2 - 3 | 412 | 64 | ||
GenBank mRNA | BC083689.1 | - | 2 - 3 | 315 | 7189368 | 64 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC083689.1, NM_001006994.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
