Assay Details
Gene Symbol: Gpr180 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006994.1 | NP_001006995.1 | 3 - 4 | 518 | 105 | |
XM_006252452.2 | XP_006252514.1 | 3 - 4 | 614 | 105 | ||
GenBank mRNA | BC083689.1 | - | 3 - 4 | 518 | 105 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
