Assay Details
Gene Symbol: Lclat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008764536.1 | XP_008762758.1 | 5 - 6 | 8231 | 73 | |
XM_008764537.1 | XP_008762759.1 | 5 - 6 | 781 | 73 | ||
XM_008764538.1 | XP_008762760.1 | 5 - 6 | 771 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
