Assay Details
Gene Symbol: Fez2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053600.2 | NP_446052.2 | 5 - 6 | 911 | 82 | |
GenBank mRNA | AB076183.1 | - | 5 - 6 | 911 | 82 | |
CB725415.1 | - | 1 - 2 | 153 | 82 | ||
U64689.1 | - | 5 - 6 | 737 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P14687 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P040705 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 840056 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 236623 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 382926 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 143911 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 942306 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 563222 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 841156 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 338755 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 121751 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 36258 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 42448 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 313972 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 164013 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 773439 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 766848 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 711500 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 407597 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 438865 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 797981 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 272487 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 700705 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 553410 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 561359 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 604800 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 529577 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 820068 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 16163 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 338392 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 393778 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 148921 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 135311 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 147800 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 72437 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 79386 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 51051 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 36284 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P040705 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P14687 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P14687 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P040705 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P14687 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P040705 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P14687 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P149098 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 57756 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 6672 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 56020 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 36285 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10887841 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887839 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887835 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887838 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887836 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887840 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887837 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887833 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887834 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887832 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10887831 | - | - |
Rat Genome 230 2.0 Array | 1388137_a_at | - | - |
Rat Genome 230 2.0 Array | 1388137_PM_a_at | - | - |
Affymetrix HT Rat Focus Array | 1388137_a_at | - | - |
Affymetrix HT Rat Focus Array | 1388137_PM_a_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0600012157.rn.1 | TC0600001722.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0600012159.rn.1 | TC0600001722.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0600012152.rn.1 | TC0600001722.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0600012158.rn.1 | TC0600001722.rn.1 | RTA TC |
Rat Expression Set 230 | 1388137_a_at | - | - |
Rat Expression Set 230 | 1388137_PM_a_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887831 | - | - |
Rat Genome U34 Set | U64689_at | - | - |
Rat Genome U34 Set | rc_AI170967_at | - | - |
Rat Genome U34 Set | U64689_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21236592 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1366361 | - | - |
RatRef-12 Expression BeadChip | ILMN_64068 | - | - |
