Assay Details
Gene Symbol: Rnaseh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013097.2 | NP_001013115.1 | 1 - 2 | 198 | 138 | |
NM_001286938.1 | NP_001273867.1 | 1 - 2 | 198 | 138 | ||
GenBank mRNA | BC091209.1 | - | 1 - 2 | 173 | 7381386 | 138 |
CB585145.1 | - | 1 - 2 | 198 | 138 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001286938.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
