Assay Details
Gene Symbol: Rnaseh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013097.2 | NP_001013115.1 | 3 - 4 | 470 | 98 | |
NM_001286938.1 | NP_001273867.1 | 3 - 4 | 470 | 98 | ||
GenBank mRNA | BC091209.1 | - | 3 - 4 | 445 | 98 | |
CB585145.1 | - | 3 - 4 | 470 | 98 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
