Assay Details
Gene Symbol: Esr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012754.1 | NP_036886.1 | 1 - 2 | 188 | 85 | |
XM_006240221.2 | XP_006240283.1 | 1 - 2 | 428 | 85 | ||
GenBank mRNA | AB530987.1 | - | 1 - 2 | 321 | 85 | |
U57439.1 | - | 1 - 2 | 188 | 85 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AB530987.1, NM_012754.1, U57439.1, XM_006240221.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
