Assay Details
Gene Symbol: Asxl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008764542.1 | XP_008762764.1 | 1 - 2 | 505 | 125 | |
XM_008764543.1 | XP_008762765.1 | 1 - 2 | 505 | 125 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
