Assay Details
Gene Symbol: Gen1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106717.1 | NP_001100187.1 | 12 - 13 | 1501 | 74 | |
XM_006239896.1 | XP_006239958.1 | 11 - 12 | 1328 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
Process(es)
double-strand break repair via homologous recombination
DNA catabolic process, endonucleolytic
regulation of centrosome duplication
resolution of recombination intermediates
resolution of mitotic recombination intermediates
positive regulation of mitotic cell cycle spindle assembly checkpoint
DNA binding
crossover junction endodeoxyribonuclease activity
double-strand break repair via homologous recombination
DNA catabolic process, endonucleolytic
regulation of centrosome duplication
resolution of recombination intermediates
resolution of mitotic recombination intermediates
positive regulation of mitotic cell cycle spindle assembly checkpoint
DNA binding
crossover junction endodeoxyribonuclease activity
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P112816 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 416292 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 213379 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 580206 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 48241 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 15412 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 861707 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 508889 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 332639 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 224933 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 34863 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 745346 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 743074 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 242104 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 260717 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 374971 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 607083 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 945780 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 419692 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 683736 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 825741 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 58397 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 87033 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 7181 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P112816 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P112816 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P112816 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P258140 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 16539 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 66430 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 17411 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10889212 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10889201 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10889203 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10889211 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10889210 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10889206 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10889205 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10889202 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10889208 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10889209 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10889200 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10889207 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10889204 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10889199 | - | - |
Rat Genome 230 2.0 Array | 1384102_at | - | - |
Rat Genome 230 2.0 Array | 1384102_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1384102_at | - | - |
Affymetrix HT Rat Focus Array | 1384102_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0600015520.rn.1 | TC0600002132.rn.1 | RTA TC |
Rat Expression Set 230 | 1384102_at | - | - |
Rat Expression Set 230 | 1384102_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10889199 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1360538 | - | - |
