Assay Details
Gene Symbol: Gen1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106717.1 | NP_001100187.1 | 9 - 10 | 1231 | 150 | |
XM_006239896.1 | XP_006239958.1 | 8 - 9 | 1058 | 150 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
