Assay Details
Gene Symbol: Ppm1g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_147209.2 | NP_671742.1 | 2 - 3 | 311 | 121 | |
GenBank mRNA | BC062083.1 | - | 2 - 3 | 311 | 121 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
