Assay Details
Gene Symbol: Slc5a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130746.1 | NP_570102.1 | 3 - 4 | 289 | 85 | |
GenBank mRNA | AF026554.1 | - | 3 - 4 | 289 | 85 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AF026554.1, NM_130746.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
