Assay Details
Gene Symbol: Gtf2a1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012136.1 | NP_001012136.1 | 2 - 3 | 144 | 76 | |
XM_006239740.2 | XP_006239802.1 | 3 - 4 | 834 | 76 | ||
XM_008764478.1 | XP_008762700.1 | 5 - 6 | 697 | 76 | ||
GenBank mRNA | BC082104.1 | - | 2 - 3 | 144 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
