Assay Details
Gene Symbol: Gtf2a1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012136.1 | NP_001012136.1 | 3 - 4 | 260 | 67 | |
XM_006239740.2 | XP_006239802.1 | 4 - 5 | 950 | 67 | ||
XM_006239741.2 | XP_006239803.1 | 3 - 4 | 849 | 67 | ||
XM_008764478.1 | XP_008762700.1 | 6 - 7 | 813 | 67 | ||
GenBank mRNA | BC082104.1 | - | 3 - 4 | 260 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
